EMAIL THIS PAGE TO A FRIEND
Journal of natural products
Phenalenone-type compounds from Musa acuminata var. "Yangambi km 5" (AAA) and their activity against Mycosphaerella fijiensis.
PMID 17428093
Abstract
Two perinaphthenone-type compounds (1 and 2) were isolated together with four known phenylphenalenones (3-6) from the rhizomes of Musa acuminata var. "Yangambi km 5". The structures of the new phenalenones were assigned as 2-hydroxy-1H-phenalen-1-one (1) and 2-methoxy-1H-phenalen-1-one (2) on the basis of their spectroscopic data and were confirmed by synthesis. Compounds 1 and 2 displayed significantly enhanced activity against Mycosphaerella fijiensis in comparison with other phenylphenalenones.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P10801
Perinaphthenone, 97%
C13H8O