EMAIL THIS PAGE TO A FRIEND
Journal of immunological methods
Use of antigen mimics to produce specific antibodies to anti-coccidial drugs.
PMID 17433817
Abstract
A range of polyclonal antibodies was successfully produced to the coccidiostatic drugs diclazuril and robenidine. Initial attempts to make immunogenic complexes of both drugs were ineffective due to difficulties encountered while trying to couple the compounds to large carrier proteins. Structural mimics, which could act as haptens for each drug, were sought and identified. The compounds identified were more open to chemical manipulation and were conjugated to carrier proteins to produce effective immunogens. The most sensitive antisera produced displayed IC50s of 1.5 ng/ml and 13 ng/ml for diclazuril and robenidine respectively. The antibody for diclazuril was shown to be specific, cross-reacting only with clazuril by 15%. The robenidine antibody displayed a low cross-reactivity of 1.2% to the compound used to produce the antibody.