EMAIL THIS PAGE TO A FRIEND
Revista brasileira de psiquiatria (Sao Paulo, Brazil : 1999)
Comments on "Weight gain management in patients with schizophrenia during treatment with olanzapine in association with nizatidine".
PMID 17435940