EMAIL THIS PAGE TO A FRIEND
Revista medico-chirurgicala a Societatii de Medici si Naturalisti din Iasi
[New biological active derivatives of indomethacin and acetylsalicylic acid. Synthesis, physico-chemical characterisation and structure validation].
PMID 17438920
Abstract
It is well known that niflumic acid glycinamide has a good antiinflammatory action useful in gum inflammatory diseases. The objective of this study was to obtain new glycinamides of acetylsalicylic acid and indomethacin, which could have a better antiinflammatory action than niflumic acid glycinamide. The study presents the synthesis, physico-chemical characterisation and structure validation of these glycinamides.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G6104
Glycinamide hydrochloride, 98%
C2H6N2O · HCl
50070
Glycinamide hydrochloride, ≥99.0% (AT)
C2H6N2O · HCl