EMAIL THIS PAGE TO A FRIEND
Basic & clinical pharmacology & toxicology
Modulation of ERG channels by XE991.
PMID 17448117
Abstract
In neuronal tissue, KCNQ2-5 channels conduct the physiologically important M-current. In some neurones, the M-current may in addition be conducted partly by ERG potassium channels, which have widely overlapping expression with the KCNQ channel subunits. XE991 and linopiridine are known to be standard KCNQ potassium channel blockers. These compounds have been used in many different tissues as specific pharmacological tools to discern native currents conducted by KCNQ channels from other potassium currents. In this article, we demonstrate that ERG1-2 channels are also reversibly inhibited by XE991 in the micromolar range (EC(50) 107 microM for ERG1). The effect has been characterized in Xenopus laevis oocytes expressing ERG1-2 and in the mammalian HEK293 cell line stably expressing ERG1 channels. The IC(50) values for block of KCNQ channels by XE991 range 1-65 microM. In conclusion, great care should be taken when choosing the concentration of XE991 to use for experiments on native potassium channels or animal studies in order to be able to conclude on selective KCNQ channel-mediated effects.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
430676
2,2,5,7,8-Pentamethyl-6-chromanol, 97%
C14H20O2
L134
Linopirdine, ≥98% (HPLC)
C26H21N3O