EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Squalene-derived flexible linkers for bioactive peptides.
PMID 17448660
Abstract
A regiochemical and stereochemical mixture of flexible linkers bearing terminal azide functionality was synthesized in two steps from squalene and was used to connect two high affinity NDP-alpha-MSH ligands or two low affinity MSH(4) ligands. The ligands were N-terminally acylated using N-hydroxysuccinimidoyl 5-hexynoate and were subsequently attached to the linker via copper-catalyzed 'click' 3+2 cyclization of the azide and alkyne moieties. In vitro biological evaluations showed that the binding affinity to the human melanocortin 4 receptor was not diminished for most linker-ligand combinations relative to the corresponding parental ligand. Statistical and cooperative binding effects were observed for dimeric constructs containing the low affinity ligand MSH(4), but not for dimeric NDP-alpha-MSH constructs, presumably due to slow off rates for this high affinity ligand.