EMAIL THIS PAGE TO A FRIEND
Journal of nanoscience and nanotechnology
Multi-walled carbon nanotube-based glucose/O2 biofuel cell with glucose oxidase and laccase as biocatalysts.
PMID 17450935
Abstract
This study describes a multi-walled carbon nanotube-based glucose/O2 biofuel cell with glucose oxidase and laccase as the anodic and cathodic biocatalysts, respectively. Upon being functionalized with L-a-phosphatidylcholine, one kind of lipid, multi-walled carbon nanotubes can serve as a support for glucose oxidase to form a three-dimensional, conducting and uniform bioanode that possesses a good bioelectrocatalytic activity toward the oxidation of glucose biofuel with solution-phased ferrocene monocarboxylic acid as the mediator to shuttle the electron transfer between glucose oxidase and multi-walled carbon nanotubes. In a similar way, the lipid-functionalized multiwalled carbon nanotubes can also be used to support the cathodic biocatalyst, i.e., laccase, and, more remarkably, to facilitate the direct electron transfer of laccase. As a result, the prepared biocathode is very active toward the reduction of oxygen without using any electron-transfer mediators. The biofuel cell has a 0.45 V open circuit potential and 34 microA/cm2 short circuit current density in phosphate buffer (pH 6.0) separated with Nafion-117 membrane with anodic compartment containing 15 mM glucose and 2 mM ferrocene monocarboxylic acid and with cathodic compartment being saturated with O2 at room temperature. A maximum power density of 3.2 microW/cm2 is obtained with ca. 0.2 V potential output.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
46264
Ferrocenecarboxylic acid, purum, ≥97.0% (Fe)
C11H10FeO2
106887
Ferrocenecarboxylic acid, 97%
C11H10FeO2