EMAIL THIS PAGE TO A FRIEND
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine
Radiolabeling of multimeric neurotensin(8-13) analogs with the short-lived positron emitter fluorine-18.
PMID 17451963
Abstract
Three methods for (18)F-labeling of dimeric and tetrameric neurotensin(8-13) derivatives were evaluated with respect to the labeling yield and the required peptide amounts. Labeling using N-succinimidyl-4-[(18)F]fluorobenzoate ([(18)F]SFB) gave low radiochemical yield for the dimeric peptides. Coupling of the tetramer with [(18)F]SFB was not successful. High yields were obtained for labeling of the aminooxy-functionalized neurotensin(8-13) dimer using 4-[(18)F]fluorobenzaldehyde ([(18)F]FBA) whilst coupling of the corresponding tetramer gave only low yields. Labeling of sulfydryl-functionalized neurotensin(8-13) derivatives using the maleinimide 4-[(18)F]fluorobenzaldehyde-O-[6-(2,5-dioxo-2,5-dihydro-pyrrol-1-yl)-hexyl]-oxime ([(18)F]FBAM) resulted in high radiochemical yields for both, the dimer and the tetramer. Therefore, [(18)F]FBAM seems to be the most suitable (18)F-labeling agent for multivalent neurotensin(8-13) derivatives.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N5266
Neurotensin Fragment 8-13 acetate salt, ≥97% (HPLC)
C38H64N12O8