EMAIL THIS PAGE TO A FRIEND
Critical reviews in toxicology
Critical observations on the neurotoxicity of silver.
PMID 17453933
Abstract
Silver is a xenobiotic element with no recognized trace metal value in the human body. It is absorbed into the body through the lungs, gastrointestinal tract, mucus membranes of the urinogenital tract, and through the skin, mainly in the form of silver protein complexes. Although silver is metabolized throughout the soft tissues, available evidence from experimental animal studies and human clinical reports has failed to unequivocally establish that it enters tissues of the central nervous system or is a cause of neurotoxic damage. Argyria characterized by deposition of particles of silver sulfide or silver selenide is the principle contraindication for using silver in medical devices or occupationally. This presents discoloration of the skin but is not regarded as a health risk or manifestation of toxicity. No evidence is available to demonstrate the toxic risk of silver to the peripheral nervous system, although silver sulfide deposits have been identified in the region of cutaneous nerves. Transitory silver sulfide deposits seen in the tissues of the blood-brain and blood-CSF barriers are mostly lysosomally bound or deposited on basement membranes or collagen without toxic effect. Silver is mostly excreted from the body in the urine and feces. Further research is indicated to evaluate the role of metal binding proteins including metallothioneins as cytoprotectants for neurological tissue.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
85131
Silver, colloidal, 65-75% Ag basis
Ag
373249
Silver, evaporation slug, diam. × L 0.6 cm × 1.2 cm, 99.99% trace metals basis
Ag
327077
Silver, flakes, 10 μm, ≥99.9% trace metals basis
Ag
326976
Silver, foil, thickness 2.0 mm, 99.9% trace metals basis
Ag
348724
Silver, foil, thickness 1.5 mm, 99.9% trace metals basis
Ag
369438
Silver, foil, thickness 1.0 mm, 99.99% trace metals basis
Ag
265543
Silver, foil, thickness 1.0 mm, 99.9% trace metals basis
Ag
265535
Silver, foil, thickness 0.5 mm, 99.99% trace metals basis
Ag
345075
Silver, foil, thickness 0.5 mm, 99.9% trace metals basis
Ag
348716
Silver, foil, thickness 0.25 mm, 99.99% trace metals basis
Ag
326984
Silver, foil, thickness 0.25 mm, 99.9% trace metals basis
Ag
265527
Silver, foil, thickness 0.1 mm, 99.9% trace metals basis
Ag
326992
Silver, foil, thickness 0.075 mm, 99.9% trace metals basis
Ag
265519
Silver, foil, thickness 0.025 mm, 99.9% trace metals basis
Ag
303372
Silver, granular, >250 μm, 99.99% trace metals basis
Ag
461032
Silver, needles, 250-600 μm, ≥99.99% trace metals basis
Ag
265500
Silver, powder, <250 μm, 99.99% trace metals basis
Ag
327107
Silver, powder, <45 μm, ≥99.99% trace metals basis
Ag
327093
Silver, powder, 5-8 μm, ≥99.9% trace metals basis
Ag
327085
Silver, powder, 2-3.5 μm, ≥99.9% trace metals basis
Ag
484059
Silver, nanopowder, <150 nm particle size, 99% trace metals basis
Ag
576832
Silver, nanopowder, <100 nm particle size, contains PVP as dispersant, 99.5% trace metals basis
Ag
265624
Silver, rod, diam. 7.0 mm, 99.99% trace metals basis
Ag
348775
Silver, rod, diam. 3.2 mm, 99.98% trace metals basis
Ag
204366
Silver, shot, 1-3 mm, 99.999% trace metals basis
Ag
327050
Silver, shot, 1-3 mm, ≥99.99% trace metals basis
Ag
267430
Silver, wire, diam. 2.0 mm, ≥99.99% trace metals basis
Ag
327018
Silver, wire, diam. 2.0 mm, 99.9% trace metals basis
Ag
348759
Silver, wire, diam. 1.5 mm, ≥99.99% trace metals basis
Ag
265594
Silver, wire, diam. 1.0 mm, ≥99.99% trace metals basis
Ag