EMAIL THIS PAGE TO A FRIEND
Journal of forensic sciences
Enhancement of ninhydrin- or DFO-treated latent fingerprints on thermal paper.
PMID 17456091
Abstract
A new method for enhancement of ninhydrin or 1,8-diazafluoren-9-one (DFO)-treated latent fingerprints on thermal paper will be described. Most thermosensitive surfaces of thermal paper become dark when treated with DFO or ninhydrin petroleum ether (NPB) solution. This effect minimizes contrast between the developed fingerprints and the background. The new method described reduces this dark staining without removing the thermosensitive layer and parts of the developed fingerprints, as occurs with acetone washing. Through the new method, the developed fingerprints appear in sharp lines and high contrast. Extensive tests were performed, leading to an optimized working solution, which charges the paper with a minimum of chemicals, is cheap, and enables a large quantity of papers to be treated in a short time. The working solution contains commercially available, nonvolatile, nitrogenous organic compounds and can be used like the application of NPB solution by dipping.