EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Genotoxicity screening for N-nitroso compounds. Electrochemical and electrochemiluminescent detection of human enzyme-generated DNA damage from N-nitrosopyrrolidine.
PMID 17457417
Abstract
We report for the first time voltammetric/electrochemiluminescent sensors applied to predict genotoxicity of N-nitroso compounds bioactivated by human cytochrome P450 enzymes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
158240
1-Nitrosopyrrolidine, 99%
C4H8N2O