EMAIL THIS PAGE TO A FRIEND
Macromolecular bioscience
Synthesis and erosion properties of PEG-containing polyanhydrides.
PMID 17457940
Abstract
To tailor the erosion rate of polyanhydrides while retaining their surface erosion characteristics, new three-component polyanhydrides of sebacic acid, 1,3-bis(p-carboxyphenoxy)propane and poly(ethylene glycol) were synthesized. The hydrophilicity of the polymer increased and its mechanical strength decreased with increasing PEG content. Correspondingly, the erosion rate increases with increasing PEG content, whereas it decreases with increasing specimen thickness. This indicates that the incorporation of poly(ethylene glycol) into traditional two-component polyanhydrides retains their surface erosion properties while making the erosion rate tunable. The new polyanhydrides hold potential for drug delivery applications.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
655538
1,3-Bis(4-carboxyphenoxy)propane, 97%
C17H16O6