EMAIL THIS PAGE TO A FRIEND
Anesteziologiia i reanimatologiia
[Midazolam in children. Alternative routes of administration].
PMID 17460996
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M-908
Midazolam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material
C18H13ClFN3