EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Steric and electronic factors influencing recognition by a simple, charge neutral norbornene based anion receptor.
PMID 17464401
Abstract
Based on (1)H NMR studies, subtle electronic factors rather than pre-organisation dictate the binding stoichiometry of the new, norbornene based, anion hosts 1 and 2 with acetate, however, the binding of dihydrogenphosphate appears to be based solely on steric constraints.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
238198
Dimethyl maleate, 96%
C6H8O4