EMAIL THIS PAGE TO A FRIEND
Journal of photochemistry and photobiology. B, Biology
Photophysical and photochemical studies of chlorophyll a and cobalt(II)tetraphenylporphyrin in poly(L-glutamate)-decylammonium ion complex.
PMID 17468007
Abstract
Spectroscopic studies were carried out on chlorophyll a and cobalt(II)tetraphenylporphyrin solubilized in a poly(L-glutamate) (Poly(Glu))-decylammonium chloride (DeAC) complex system, in the presence of methylviologen (MV2+). The cooperative binding occurred between the anionic Poly(Glu) and the cationic DeAC, leading to the formation of micelle-like hydrophobic clusters of DeAC and also the change in conformation of the Poly(Glu) from the random coil to the alpha-helix. All of the absorption spectra, the fluorescence quantum yields and the fluorescence lifetimes indicated the existence of equilibrium between the aggregated biofunctional molecules in the bulk phase and the monomeric species in the complex phase of the Poly(Glu)-DeAC solution. The fluorescence quenching of the biofunctional molecules by methylviologen indicates that the conformation-dependent electron transfer occurs in the complex phase.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
30692
Decylamine, ≥99.0% (GC)
C10H23N
D2404
Decylamine, 95%
C10H23N
442540
Decylamine, analytical standard
C10H23N