EMAIL THIS PAGE TO A FRIEND
Bioresource technology
Kinetics and thermodynamics of the adsorption of some dyestuffs from aqueous solution by poplar sawdust.
PMID 17470390
Abstract
The effect of temperature on the adsorption of metanil yellow (MY) (acidic) and methylene blue (MB) (basic) by poplar sawdust was investigated. In addition, the amounts of NaHCO(3), Na(2)CO(3), NaOH and C(2)H(5)ONa adsorbed by 1g of poplar sawdust to determine its surface acidity were also determined. Kinetical data obtained at different temperatures (293 K, 313 K and 333 K) for the adsorption of each dyestuff by poplar sawdust were applied to the pseudo first-order, the pseudo second-order and the intraparticle diffusion equations, and the rate constants of first-order adsorption (k(1)), the rate constants of second-order adsorption (k(2)) and intraparticle diffusion rate constants (k(p)) at these temperatures were calculated, respectively. In addition, isothermal data obtained at different temperatures (293 K, 313 K and 333 K) for the adsorption of each dyestuff by poplar sawdust were applied to thermodynamical equations, and thermodynamical parameters (DeltaG, DeltaH and DeltaS) were also calculated.