EMAIL THIS PAGE TO A FRIEND
Nucleic acids research
Transcription-coupled deposition of histone modifications during MHC class II gene activation.
PMID 17478518
Abstract
Posttranslational histone modifications associated with actively expressed genes are generally believed to be introduced primarily by histone-modifying enzymes that are recruited by transcription factors or their associated co-activators. We have performed a comprehensive spatial and temporal analyses of the histone modifications that are deposited upon activation of the MHC class II gene HLA-DRA by the co-activator CIITA. We find that transcription-associated histone modifications are introduced during two sequential phases. The first phase precedes transcription initiation and is characterized exclusively by a rapid increase in histone H4 acetylation over a large upstream domain. All other modifications examined, including the acetylation and methylation of several residues in histone H3, are restricted to short regions situated at or within the 5' end of the gene and are established during a second phase that is concomitant with ongoing transcription. This second phase is completely abrogated when elongation by RNA polymerase II is blocked. These results provide strong evidence that transcription elongation can play a decisive role in the deposition of histone modification patterns associated with inducible gene activation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
06-599
Anti-acetyl-Histone H3 Antibody, Detect acetyl-Histone H3 with Anti-acetyl-Histone H3 Antibody (Rabbit Polyclonal Antibody), that has been shown to work in WB, ICC, ChIP, ChIP-seq.
07-353
Anti-acetyl-Histone H3 (Lys14) Antibody, Anti-acetyl-Histone H3 (Lys14) Antibody is a rabbit polyclonal antibody for detection of acetyl-Histone H3 (Lys14) also known as H3K14Ac, Histone H3 (acetyl K14) & has been validated in ChIP, WB, ChIP-seq, DB, Mplex.
07-355
Anti-acetyl-Histone H3 (Lys23) Antibody, Use Anti-acetyl-Histone H3 (Lys23) Antibody (Rabbit Polyclonal Antibody) validated in ChIP, WB to detect acetyl-Histone H3 (Lys23) also known as H3K23Ac, Histone H3 (acetyl K23).
07-360
Anti-acetyl-Histone H3 (Lys27) Antibody, Anti-acetyl-Histone H3 (Lys27) Antibody is a rabbit polyclonal antibody for detection of Histone H3 acetylated on lysine 27. Also known as Anti-H3K27ac this antibody is published in peer reviewed journals and is specificity verified by dot blot (DB) and validated in ChIP, ChIP-seq, WB, DB, Mplex.
07-352
Anti-acetyl-Histone H3 (Lys9) Antibody, Anti-acetyl-Histone H3 (Lys9) Antibody is a Rabbit Polyclonal Antibody for detection of acetyl-Histone H3 (Lys9) also known as H3K9Ac, Histone H3 (acetyl K9) and has been published and validated in ChIP, WB, Mplex.
06-866
Anti-acetyl-Histone H4 Antibody, Use Anti-acetyl-Histone H4 Antibody (Rabbit Polyclonal Antibody) has been published and validated in ChIP, WB, ICC to detect acetyl-Histone H4 also known as H4 histone family member A, histone 1-H4a.
07-595
Anti-acetyl-Histone H4 (Lys12) Antibody, Use Anti-acetyl-Histone H4 (Lys12) Antibody (Rabbit Polyclonal Antibody) validated in ChIP, DB, WB, ChIP-seq to detect acetyl-Histone H4 (Lys12) also known as H4K12Ac, Histone H4 (acetyl K12).
07-329
Anti-acetyl-Histone H4 (Lys16) Antibody, Anti-acetyl-Histone H4 (Lys16) Antibody is a rabbit polyclonal antibody for detection of acetyl-Histone H4 (Lys16) also known as H4K16Ac, Histone H4 (acetyl K16 & has been published and validated in ChIP, WB, Mplex, PIA, DB, ChIP-seq.
07-328
Anti-acetyl-Histone H4 (Lys8) Antibody, Use Anti-acetyl-Histone H4 (Lys8) Antibody (Rabbit Polyclonal Antibody) validated in ChIP, IP, WB, DB, ChIP-seq to detect acetyl-Histone H4 (Lys8) also known as H4K8Ac, Histone H4 (acetyl K8).
07-214
Anti-dimethyl-Histone H3 (Arg17) Antibody, Anti-dimethyl-Histone H3 (Arg17) Antibody is a Rabbit Polyclonal Antibody for detection of dimethyl-Histone H3 (Arg17) also known as H3R17me2, Histone H3 (di methyl R17) & has been validated in ELISA, ICC peptide inhibition & WB.