Experimental neurology
Antidepressants inhibit interferon-gamma-induced microglial production of IL-6 and nitric oxide.
PMID 17481608
Circumstantial evidence has suggested that activated microglia may be associated with the pathogenesis of depression. Pro-inflammatory cytokines may also be involved. Therefore, we examined the effects of various types of antidepressants, as well as the mood-stabilizer lithium chloride, on interferon-gamma (IFN-gamma)-induced microglial production of the pro-inflammatory mediators interleukin-6 (IL-6) and nitric oxide (NO). Treatment of the murine microglial 6-3 cells with 100 U/ml of IFN-gamma resulted in an eightfold increase in IL-6 and a tenfold increase in NO into the culture medium. Pretreatment with the selective serotonin reuptake inhibitor fluvoxamine, the relatively selective noradrenaline reuptake inhibitor reboxetine, or the non-selective monoaminergic reuptake inhibitor imipramine, significantly inhibited IL-6 and NO production in a dose-dependent manner. These inhibitions were reversed significantly by SQ 22536, a cyclic adenosine monophosphate (cAMP) inhibitor, and, except for reboxetine, by the protein kinase A (PKA) inhibitor Rp-adenosine3',5'-cyclic monophosphorothioate triethylammonium salt (Rp-3',5'-cAMPS). Lithium chloride, which is believed to act by inhibiting the calcium-dependent release of noradrenaline, had a different spectrum of action on microglial 6-3 cells. It enhanced IFN-gamma-stimulated IL-6 production and inhibited NO production. The inhibitory effect of lithium chloride was not reversed by either SQ 22536 or Rp-3',5'-cAMPS. These results suggest that antidepressants have inhibitory effects on IFN-gamma-activated microglia and these effects are, at least partially, mediated by the cAMP-dependent PKA pathway. On the other hand, the mood stabilizer and anti-manic agent lithium chloride has mixed effects on IFN-gamma-induced microglial activation.
Related Materials
Product #
Molecular Formula
Add to Cart
Adenosine 5′-(β,γ-imido)triphosphate lithium salt hydrate, ≥93% (HPLC), powder
C10H17N6O12P3 · xLi+ · yH2O