EMAIL THIS PAGE TO A FRIEND
Physical chemistry chemical physics : PCCP
266 nm photolysis of CF3I and C2F5I studied by diode laser gain FM spectroscopy.
PMID 17487320
Abstract
Frequency modulated diode laser based absorption at 1.315 microm has been used to measure the Doppler lineshapes of the I((2)P(1/2)-(2)P(3/2)) transition in atomic iodine produced from the 266 nm photolysis of both CF(3)I and C(2)F(5)I. Wavelength resolved laser gain is seen following photolysis as excited iodine atoms ((2)P(1/2)) are produced with a quantum yield close to unity from photolysis of both parent molecules. Time resolved measurements were made and the nascent speed distribution and translational anisotropy parameter, beta were determined. Mean atomic speeds of 800 and 850 ms(-1), which correspond to 83 and 68% of the maximum possible kinetic energy release into the iodine photofragment, were determined for photolysis of CF(3)I and C(2)F(5)I, respectively. The nascent translational anisotropy parameter was found to be beta = 1.77 +/- 0.05 for CF(3)I and beta = 1.69 +/- 0.05 for C(2)F(5)I. These values are explicable in terms of parent rotational motion and non-adiabatic processes in the exit channel.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
171441
Trifluoroiodomethane, 99%
CF3I