EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Mitotic regulation of SIRT2 by cyclin-dependent kinase 1-dependent phosphorylation.
PMID 17488717
Abstract
Sirtuins are evolutionarily conserved NAD(+)-dependent deacetylases and ADP-ribosyltransferases involved in the regulation of cell division, apoptosis, DNA damage repair, genomic silencing, and longevity. Recent studies have focused on identifying target substrates for human sirtuin enzymatic activity, but little is known about processes that directly regulate their function. Here, we demonstrate that SIRT2 is phosphorylated both in vitro and in vivo on serine 368 by the cell-cycle regulator, cyclin-dependent kinase 1, and dephosphorylated by the phosphatases CDC14A and CDC14B. Overexpression of SIRT2 mediates a delay in cellular proliferation that is dependent on serine 368 phosphorylation. Furthermore, mutation of serine 368 reduces hyperploidy in cells under mitotic stress due to microtubule poisons.