EMAIL THIS PAGE TO A FRIEND
Chembiochem : a European journal of chemical biology
Straightforward preparation and assay of aspartyl protease substrates with an internal thioester linkage.
PMID 17492698
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
143057
Aldrithiol-4, 98%
C10H8N2S2