EMAIL THIS PAGE TO A FRIEND
Neuropharmacology
Role of G(q) protein in behavioral effects of the hallucinogenic drug 1-(2,5-dimethoxy-4-iodophenyl)-2-aminopropane.
PMID 17493641
Abstract
Extensive evidence suggests that 5-HT2 receptors may play a role in mental disorders including schizophrenia. In addition, several studies indicate that G(q)-coupled 5-HT(2A) receptors are likely targets for the initiation of events leading to the hallucinogenic behavior elicited by lysergic acid diethylamide (LSD), (+/-)1-(2,5-dimethoxy-4-iodophenyl)-2-aminopropane (DOI), and related drugs. However, 5-HT(2A) receptors couple to other G proteins in addition to G(q) protein. To evaluate the role of the G(q) signaling pathway in DOI-induced behaviors, we utilized two behavioral models of 5-HT(2A) receptor activation: induction of head-twitches by DOI, a common response to hallucinogenic drugs in rodents, and DOI elicited anxiolytic-like effects in the elevated plus maze. Experimental subjects were genetically modified mice [Galpha(q)(-/-)] in which the G(q) alpha gene was eliminated. Galpha(q)(-/-) mice exhibited a decrease in DOI-induced head-twitches, when compared to wild-type littermates. In addition, the DOI-induced increase in anxiolytic-like behavior was abolished in Galpha(q)(-/-) mice. These results, combined with our finding that DOI-induced FOS expression in the medial prefrontal cortex was also eliminated in Galpha(q)(-/-) mice, suggests a key role for G(q) protein in hallucinogenic drug effects.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I5763
Indophenol
C12H9NO2