EMAIL THIS PAGE TO A FRIEND
Cellular and molecular life sciences : CMLS
Human and yeast zeta-crystallins bind AU-rich elements in RNA.
PMID 17497241
Abstract
Zeta-crystallins constitute a family of proteins with NADPH:quinone reductase activity found initially in mammalian lenses but now known to be present in many other organisms and tissues. Few proteins from this family have been characterized, and their function remains unclear. In the present work, zeta-crystallins from human and yeast (Zta1p) were expressed, purified and characterized. Both enzymes are able to reduce ortho-quinones in the presence of NADPH but are not active with 2-alkenals. Deletion of the ZTA1 gene makes yeast more sensitive to menadione and hydrogen peroxide, suggesting a role in the oxidative stress response. The human and yeast enzymes specifically bind to adenine-uracil rich elements (ARE) in RNA, indicating that both enzymes are ARE-binding proteins and that this property has been conserved in zeta-crystallins throughout evolution. This supports a role for zeta-crystallins as trans-acting factors that could regulate the turnover of certain mRNAs.