EMAIL THIS PAGE TO A FRIEND
Zhonghua lao dong wei sheng zhi ye bing za zhi = Zhonghua laodong weisheng zhiyebing zazhi = Chinese journal of industrial hygiene and occupational diseases
[Involvement of Caspase-3 and p38 mitogen-activated protein kinase in MMT-induced apoptosis in PC-3M cells].
PMID 17498346
Abstract
To explore the function of Caspase-3 and p38 MAPK in MMT-induced apoptosis in PC-3M cells. After incubation of PC-3M cells with 1 mmol/L MMT, the activity of Caspase-3 was examined. The influence on cells viability of Z-DEVD-FMK, a Caspase-3-specific peptide inhibitor, was also examined. Western blot was used to examine the change of p38 MAPK. The effect on cells viability and Caspase-3 activity of SB203580, a specific inhibitor of p38 MAPK, were also examined. The activity of Caspase-3 increased significantly in MMT-induced apoptosis in PC-3M cells /9P < 0.01), and Z-DEVD-FMK could protect cells from apoptosis (P < 0.01). In this course, the phosphorylation of p38 MAPK could be observed. SB203580 inhibited Caspase-3 activity (P < 0.05) and prevented PC-3M cells from MMT-induced apoptosis (P < 0.05). Caspase-3 and p38 MAPK are involved in MMT-induced PC-3M cells apoptosis.