EMAIL THIS PAGE TO A FRIEND
Journal of clinical laboratory analysis
Improved method to calculate the antibody avidity index.
PMID 17506479
Abstract
The methods currently used to determine the immunoglobulin avidity index (AI) require the choice of a reference point in the ELISA titration curve. Since both curves, with and without denaturating reagents, seldom run in parallel, the AI value becomes highly dependent on this reference. The new method for AI calculation presented here takes into account the whole data of the ELISA titration curve in which the final numerical AI is the average of each point.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
12669
Potassium thiosulfate, ≥95%
K2O3S2