EMAIL THIS PAGE TO A FRIEND
Immunity
Initiation of plasma-cell differentiation is independent of the transcription factor Blimp-1.
PMID 17509907
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
05-1573
Anti-Pax5 Antibody, clone 1H9, clone 1H9, from rat