EMAIL THIS PAGE TO A FRIEND
American journal of clinical pathology
NK-cell lymphomas and leukemias: a spectrum of tumors with variable manifestations and immunophenotype.
PMID 17509983
Abstract
Natural killer (NK) cells are lymphocytes that have some phenotypic and functional similarities to cytotoxic T cells but do not express the T-cell receptor complex. NK-cell malignancies may be localized or disseminated at initial examination, and most behave aggressively. The variable presentation of NK-cell lymphomas and leukemias suggests that they represent a spectrum of disease, with Epstein-Barr virus (EBV) implicated in the pathogenesis of most cases. Using cases presented in Session 10 of the 2005 Society for Hematopathology/European Association for Haematopathology Workshop on T-cell and NK-cell malignancies, we discuss outstanding issues in the classification and diagnosis of NK-cell malignancies. These difficulties are related to unusual sites of manifestation, atypical immunophenotypic features, and EBV+ T-cell tumors that resemble classical extranodal NK/T-cell lymphoma, nasal-type (EN-NK/T-NT). Although some of these cases can be grouped into ENNK/T-NT, classification of tumors that have atypical or discordant features will remain controversial, particularly when EBV is absent.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
381R-1
TIA-1 (EP243) Rabbit Monoclonal Antibody