EMAIL THIS PAGE TO A FRIEND
Vision research
Oscillatory potentials of the slow-sequence multifocal ERG in primates extracted using the Matching Pursuit method.
PMID 17512027
Abstract
This study used the Matching Pursuit (MP) method, a time-frequency analysis, to identify and characterize oscillatory potentials (OPs) in the primate electroretinogram (ERG). When the slow-sequence mfERG from the macular region of the retina was matched with Gabor functions, OPs were identified in two distinct bands: a high-frequency band peaking around 150 Hz that contributes to early OPs, and a low-frequency band peaking around 80 Hz that contributes to both early and late OPs. Pharmacological blockade and experimental glaucoma studies showed that the high-frequency OPs depend upon sodium-dependent spiking activity of retinal ganglion cells, whereas the low-frequency OPs depend primarily upon non-spiking activity of amacrine cells, and more distal retinal activity.