EMAIL THIS PAGE TO A FRIEND
Disaster management & response : DMR : an official publication of the Emergency Nurses Association
A mass casualty incident involving children and chemical decontamination.
PMID 17517363
Abstract
Mass casualty incidents involving contaminated children are a rare but ever-present possibility. In this article we outline one such event that resulted in 53 pediatric patients and 3 adults presenting to the emergency department of a children's hospital for decontamination and treatment. We pay special attention to the training that allowed this responses to occur. We also outline the institutional response with emphasis on incident command, communication, and resource utilization. Specific lessons learned are explored in detail. Finally, we set forth a series of recommendations to assist other institutions should they be called upon to care for and decontaminate pediatric patients.
Related Materials