EMAIL THIS PAGE TO A FRIEND
American journal of physiology. Regulatory, integrative and comparative physiology
3,5-Diiodothyronine in vivo maintains euthyroidal expression of type 2 iodothyronine deiodinase, growth hormone, and thyroid hormone receptor beta1 in the killifish.
PMID 17522123
Abstract
Until recently, 3,5-diiodothyronine (3,5-T(2)) has been considered an inactive by-product of triiodothyronine (T(3)) deiodination. However, studies from several laboratories have shown that 3,5-T(2) has specific, nongenomic effects on mitochondrial oxidative capacity and respiration rate that are distinct from those due to T(3). Nevertheless, little is known about the putative genomic effects of 3,5-T(2). We have previously shown that hyperthyroidism induced by supraphysiological doses of 3,5-T(2) inhibits hepatic iodothyronine deiodinase type 2 (D2) activity and lowers mRNA levels in the killifish in the same manner as T(3) and T(4), suggesting a pretranslational effect of 3,5-T(2) (Garcia-G C, Jeziorski MC, Valverde-R C, Orozco A. Gen Comp Endocrinol 135: 201-209, 2004). The question remains as to whether 3,5-T(2) would have effects under conditions similar to those that are physiological for T(3). To this end, intact killifish were rendered hypothyroid by administering methimazole. Groups of hypothyroid animals simultaneously received 30 nM of either T(3), reverse T(3), or 3,5-T(2). Under these conditions, we expected that, if it were bioactive, 3,5-T(2) would mimic T(3) and thus reverse the compensatory upregulation of D2 and tyroid receptor beta1 and downregulation of growth hormone that characterize hypothyroidism. Our results demonstrate that 3,5-T(2) is indeed bioactive, reversing both hepatic D2 and growth hormone responses during a hypothyroidal state. Furthermore, we observed that 3,5-T(2) and T(3) recruit two distinct populations of transcription factors to typical palindromic and DR4 thyroid hormone response elements. Taken together, these results add further evidence to support the notion that 3,5-T(2) is a bioactive iodothyronine.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D0629
3,5-Diiodo-L-thyronine, thyroid hormone analog
C15H13I2NO4