EMAIL THIS PAGE TO A FRIEND
Xenobiotica; the fate of foreign compounds in biological systems
Disposition of sanguinarine in the rat.
PMID 17523056
Abstract
Sanguinarine is an alkaloid with known antibiotic and anti-inflammatory activity and its pharmacokinetics have been studied in the rat after a single oral dose (10 mg kg(-1) body weight). Alkaloid determination in the plasma and liver was carried out by high-performance liquid chromatography-electrospray ionization mass spectrometry (HPLC/ESI-MS). The pharmacokinetic parameters (t(max), c(max), AUC(0-->t) and AUC(0-->infinity)) were determined for sanguinarine and dihydrosanguinarine, the major components detected in plasma. The first step in sanguinarine metabolism in the rat was the reduction of the iminium bond resulting in formation of the less toxic dihydrosanguinarine. Both compounds were completely eliminated from the plasma and liver after 24 h and not detected in urine. After a single oral dose of (3)H-sanguinarine, more than 42% of the ingested radioactivity was present in gastrointestinal tract. Benz[c]acridine, up to date the only sanguinarine metabolite referred to in the literature, was not detected in the plasma, liver or urine.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
BCR158
Benz[c]acridine, BCR® certified Reference Material
C17H11N