EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Discovery of para-alkylthiophenoxyacetic acids as a novel series of potent and selective PPARdelta agonists.
PMID 17524639
Abstract
A novel series of potent and selective PPARdelta agonists, para-alkylthiophenoxyacetic acids, was identified. The synthesis and structure-activity relationships are described.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T33006
(Phenylthio)acetic acid, 96%
C8H8O2S