EMAIL THIS PAGE TO A FRIEND
Molecular pharmacology
Structure-function studies of allosteric agonism at M2 muscarinic acetylcholine receptors.
PMID 17525129
Abstract
The M2 muscarinic acetylcholine receptor (mAChR) possesses at least one binding site for allosteric modulators that is dependent on the residues (172)EDGE(175), Tyr(177), and Thr(423). However, the contribution of these residues to actions of allosteric agonists, as opposed to modulators, is unknown. We created mutant M2 mAChRs in which the charge of the (172)EDGE(175) sequence had been neutralized and each Tyr(177) and Thr(423) was substituted with alanine. Radioligand binding experiments revealed that these mutations had a profound inhibitory effect on the prototypical modulators gallamine, alcuronium, and heptane-1,7-bis-[dimethyl-3'-phthalimidopropyl]-ammonium bromide (C7/3-phth) but minimal effects on the orthosteric antagonist [3H]N-methyl scopolamine. In contrast, the allosteric agonists 4-I-[3-chlorophenyl]carbamoyloxy)-2-butynyltrimethylammnonium chloride (McN-A-343), 4-n-butyl-1-[4-(2-methylphenyl)-4-oxo-1-butyl] piperidine hydrogen chloride (AC-42), and the novel AC-42 derivative 1-[3-(4-butyl-1-piperidinyl)propyl]-3,4-dihydro-2(1H)-quinolinone (77-LH-28-1) demonstrated an increased affinity or proportion of high-affinity sites at the combined EDGE-YT mutation, indicating a different mode of binding to the prototypical modulators. Subsequent functional assays of extracellular signal-regulated kinase (ERK)1/2 phosphorylation and guanosine 5'-(gamma-[(35)S]thio)triphosphate ([(35)S]GTPgammaS) binding revealed minimal effects of the mutations on the orthosteric agonists acetylcholine (ACh) and pilocarpine but a significant increase in the efficacy of McN-A-343 and potency of 77-LH-28-1. Additional mutagenesis experiments found that these effects were predominantly mediated by Tyr(177) and Thr(423), rather than the (172)EDGE(175) sequence. The functional interaction between each of the allosteric agonists and ACh was characterized by high negative cooperativity but was consistent with an increased allosteric agonist affinity at the combined EDGE-YT mutant M2 mAChR. This study has thus revealed a differential role of critical allosteric site residues on the binding and function of allosteric agonists versus allosteric modulators of M2 mAChRs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C7041
McN-A-343, ≥98% (TLC)
C14H18Cl2N2O2