EMAIL THIS PAGE TO A FRIEND
Georgian medical news
[Hormonal contraception and cervix of the uterus pathology].
PMID 17525492
Abstract
The effect of hormonal contraception on cervix of the uterus of young nullipara women with ectopia was investigated. Cohort study was carried out by using simple blind method. By means of statistical data manipulation correlation between the application of hormonal contraceptives and changes of colposcopy pictures in the zone of pathology was displayed. Excluton showed low percent anomalous colpscopy picture in pathology zone. Excluton was declared as medicine of choice for nullipara women with ectopia. Still it is necessary to conduct a strong dynamic control of cervix of the uterus (colposcopy and pap-smear test) in order to avoid complications.