EMAIL THIS PAGE TO A FRIEND
Internal medicine (Tokyo, Japan)
Do the effects of long-term lomerizine administration differ with age?
PMID 17527044
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L6295
Lomerizine dihydrochloride, ≥98% (HPLC), powder
C27H30F2N2O3·2HCl