EMAIL THIS PAGE TO A FRIEND
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials
[Studies on the chemical constituents from Inula cappa (II)].
PMID 17539299
Abstract
To study the chemical constituents of the root of Inula cappa. The compounds were isolated and purified by recrystallization and chromatography with silica gel column and Sephadex LH-20 column. Their structures were identified by physico-chemical properties and spectral analysis. Seven compounds were isolated as scopolin (I), octacosanoic acid (II), tritriacontane (III), (2S,3S,4R,8E)-2-[(2'R)-2'-hydroxydocosanosylamino]-octadecane-1,3,4-triol(IV),(2S,3S,4R,8E)-2-[(2'R)-2'-hydroxytricosanosylamino]-octadecane-1,3,4-triol(V), (2S,3S,4R,8E)-2-[(2'R)-2'-hydroxytetracosanosylamino]-octadecane-1,3,4-triol(VI), (2S,3S,4R,8E)-2-[(2'R)-2'-hydroxypentacosanosylamino]-octadecane-1,3,4-triol(VIII), (2S,3S,4R,8E)-2-[(2'R)-2'-hydroxyhexacosanosylamino]-octadecane-1,3,4-triol(VIII). All compounds were isolated from Inula cappa for the first time.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
284432
Octacosanoic acid, synthetic, ≥98%
C28H56O2
93435
Tritriacontane, analytical standard
C33H68