EMAIL THIS PAGE TO A FRIEND
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials
[Study on the active components of Nardostachys chinensis].
PMID 17539300
Abstract
To study on the active components of Nardostachys chinensis Batal, the compounds were isolated and purified by chromatographic methods, with their structures identified by spectral analysis and comparison with published data. 9 compounds were obtained and their structures were identified as acaciin, ursolie acid, octacosanol, kanshone A, nardosinonediol, nardosinone, aristolen-9beta-ol, oleanolic acid and beta-sitosterol. Acaciin, ursolie acid and octacosanol were obtained from Nardostachys chinensis Batal. for the first time. Acaciin and ursolie are the active components of antihiotics and anti-inflammatory.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O3379
1-Octacosanol, ≥99% (GC)
C28H58O