EMAIL THIS PAGE TO A FRIEND
Macromolecular bioscience
Lipase-catalyzed synthesis and curing of high-molecular-weight polyricinoleate.
PMID 17541925
Abstract
High-molecular-weight polyricinoleate, with an M(w) of 100,600, was enzymatically prepared by the polycondensation of methylricinoleate using immobilized lipase from Pseudomonas cepacia (IM-PC) in bulk in the presence of 4 A molecular sieves at 80 degrees C for 7 d. Polyricinoleate was a viscous liquid at room temperature with a glass transition temperature (T(g)) of -74.8 degrees C, showed no crystallinity and was biodegraded by activated sludge. Polyricinoleate was readily cured using a dicumyl peroxide at 170 degrees C for 30 min to produce a chloroform insoluble crosslinked polyricinoleate with a hardness of 50A using durometer A.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
329541
Dicumyl peroxide, 98%
C18H22O2