EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Synthesis and preliminary biological characterization of new semisynthetic derivatives of ramoplanin.
PMID 17542573
Abstract
Ramoplanin is a glycolipodepsipeptide antibiotic active against Gram-positive bacteria including vancomycin-resistant enterococci. Ramoplanin inhibits bacterial cell wall biosynthesis by a mechanism different from that of glycopeptides and hence does not show cross-resistance with these antibiotics. The systemic use of ramoplanin has been so far prevented because of its low local tolerability when injected intravenously. To overcome this problem, the fatty acid side chain of ramoplanin was selectively removed and replaced with a variety of different carboxylic acids. Many of the new ramoplanin derivatives showed antimicrobial activity similar to that of the natural precursor coupled with a significantly improved local tolerability. Among them the derivative in which the 2-methylphenylacetic acid has replaced the di-unsaturated fatty acid side chain (48) was selected as the most interesting compound and submitted to further in vitro and in vivo characterization studies.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R1781 Ramoplanin