EMAIL THIS PAGE TO A FRIEND
Cellular and molecular biology (Noisy-le-Grand, France)
Delayed 24 hours Nomega-nitro-L-arginine methyl ester injection induces pharmacological cardioprotection against reperfusion injury.
PMID 17543224
Abstract
Previous studies indicate that adenosine supplementation or nitric oxide synthase (NOS) inhibition during reperfusion exert protective effects against myocardial ischemia-reperfusion (I/R) injury. We wanted to test the hypothesis that NOS inhibition before I/R also protects the myocardium against further injury and aimed to determine the involvement of adenosine receptors in a perfused rat heart model. Rats were injected with 10 mg/kg of L-NAME (N(omega)-nitro-L-arginine methyl ester) or L-NAME + SPT (8-(p-sulfophenyl)-theophylline)--an adenosine antagonist - at 2 x 25 mg/kg or with a saline buffer, 24 hrs prior to heart excision. The hearts, perfused retrogradely were subjected to 60 min of global ischemia followed by 120 min reperfusion. L-NAME decreased NOx (nitrite and nitrate) production (16.2 +/- 3.2 vs. 7.0 +/- 1.8 micromol/L; P<0.05) in vivo and increased the release of troponin I (0.04 +/- 0.01 vs. 0.02 +/- 0.01 microg/L; P<0.05) in the plasma, compared to controls. After 120 min of reperfusion, there was a higher release of adenosine (26.1 +/- 2.2 vs. 2.4 +/- 1.2 nmol/min; P<0.01) and a decrease in troponin I levels (0.19 +/-0.07 vs. 0.59 +/- 0.16 ng/min; P<0.05) in the L-NAME group compared to controls. These results were accompanied by a higher proportion of recovery of left ventricular developed pressure (72.0 +/- 4.0 vs. 60.0 +/- 4.0%; P<0.05) and coronary flow (72.0 +/- 5.0 vs. 51.0 +/- 4.0%; P<0.05) in the L-NAME group. These beneficial effects were not blocked by the adenosine receptor antagonist. The present study reveals that L-NAME protects against I/R injury when the inhibitor is administered 24 hrs before ischemia. The beneficial effects observed in this model appear to be independent of adenosine receptor stimulation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
11030
L-Arginine methyl ester dihydrochloride, ≥98.0% (AT)
C7H16N4O2 · 2HCl