EMAIL THIS PAGE TO A FRIEND
Clinical cancer research : an official journal of the American Association for Cancer Research
KIF14 messenger RNA expression is independently prognostic for outcome in lung cancer.
PMID 17545527
Abstract
The mitotic kinesin KIF14 is overexpressed in multiple cancers including lung cancer. Therefore, we investigated KIF14 expression in association with clinical variables and the effect of KIF14 on in vitro colony formation in non-small-cell lung carcinoma. RNA was extracted from 129 untreated, resected tumors and KIF14 expression was quantified by real-time reverse transcription-PCR. Associations with clinical variables were determined by standard statistical methods. KIF14 expression was knocked down by small interfering RNA in H1299 and HeLa cells; proliferation and growth in soft agar were assayed. Squamous cell carcinoma had the highest KIF14 level, followed by large-cell undifferentiated carcinoma, then adenocarcinoma (P = 0.002). KIF14 level decreased with differentiation (P = 0.01) but was not associated with pathologic stage, T or N stage, or sex. When dichotomized about the median, KIF14 overexpression significantly decreased disease-free survival (Kaplan-Meier log-rank, P = 0.01) and trended toward decreasing overall survival (P = 0.08). In a univariate Cox proportional hazard regression, increasing KIF14 expression decreased disease-free survival [P = 0.01; hazard ratio, 1.44 (95% confidence interval, 1.09-1.91)]. In a multivariate Cox regression, including stage, differentiation, histology, and tumor purity as covariates, KIF14 overexpression remained an independent prognostic factor for disease-free survival [P = 0.01; hazard ratio, 1.45 (95% confidence interval, 1.09-1.94)]. Knockdown of KIF14 in non-small-cell lung carcinoma and cervical carcinoma cell lines decreased proliferation and colony formation in soft agar. KIF14 expression is independently prognostic for disease-free survival in lung cancer and knockdown decreases tumorigenicity in vitro, showing that it is a clinically relevant oncogene and an exciting therapeutic target for further study.