EMAIL THIS PAGE TO A FRIEND
Hippocampus
Cell cycle alteration and decreased cell proliferation in the hippocampal dentate gyrus and in the neocortical germinal matrix of fetuses with Down syndrome and in Ts65Dn mice.
PMID 17546680