EMAIL THIS PAGE TO A FRIEND
Seminars in dialysis
Recognition, treatment, and prevention of propylene glycol toxicity.
PMID 17555487
Abstract
Propylene glycol is a commonly used solvent for oral, intravenous, and topical pharmaceutical preparations. Although it is considered safe, large intravenous doses given over a short period of time can be toxic. Underlying renal insufficiency and hepatic dysfunction raise risk for toxicity. Toxic effects include hyperosmolality, increased anion gap metabolic acidosis (due to lactic acidosis), acute kidney injury, and sepsis-like syndrome. Treatment of toxicity includes hemodialysis to effectively remove propylene glycol. Prevention is best achieved by limiting the dose of propylene glycol infused.