EMAIL THIS PAGE TO A FRIEND
Journal of Chemical Neuroanatomy
Distribution and morphology of cholinergic, catecholaminergic and serotonergic neurons in the brain of Schreiber's long-fingered bat, Miniopterus schreibersii.
PMID 17560075