EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Selection of supported cobalt substrates in the presence of oxone for the oxidation of monuron.
PMID 17567139
Abstract
The immobilization of cobalt ion on different media to catalyze oxone has been investigated. A probe herbicide, Monuron, was effectively degraded by using Co2+/oxone systems. For Co2+ supported on zeolite, 100% of Monuron could be removed within a 10 min reaction time. However, the recycling of the spent Co-zeolite catalyst using various posttreatments did not give a promising result. This is likely because the zeolite particles in solution have blocked and significantly attenuated the incident UV light from reducing Co3+ to Co2+. On the contrary, the use of cationic resin has minimized these problems. In the process of Co-resin/oxone/UV, faster Monuron decay could be achieved than that in the dark reaction. In the presence of UV, a significant drop of total organic carbon (TOC) was also observed in this approach suggesting an effective and clean process for Monuron mineralization.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36174
Monuron, PESTANAL®, analytical standard
C9H11ClN2O