EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Asymmetric fluorination of alpha-aryl acetic acid derivatives with the catalytic system NiCl2-Binap/R3SiOTf/2,6-lutidine.
PMID 17568468