EMAIL THIS PAGE TO A FRIEND
The Journal of Comparative Neurology
VGLUT1 and VGLUT2 innervation in autonomic regions of intact and transected rat spinal cord.
PMID 17570127
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AG208 Vesicular Glutamate Transporter 1, control peptide for AB5905