EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Determination of urea using high-performance liquid chromatography with fluorescence detection after automated derivatisation with xanthydrol.
PMID 17570379
Abstract
A high-performance liquid chromatography (HPLC) method for the determination of urea that incorporates automated derivatisation with xanthydrol (9H-xanthen-9-ol) is described. Unlike the classic xanthydrol approach for the determination of urea, which involves the precipitation of dixanthylurea (N,N'-di-9H-xanthen-9-ylurea), the derivatisation procedure employed in this method produces N-9H-xanthen-9-ylurea, which remains in solution and can be quantified using fluorescence detection (lambda(ex)=213 nm; lambda(em)=308 nm) after chromatographic separation from interferences. The limit of detection for urea was 5 x 10(-8) M (0.003 mg L(-1)). This method was applied to the determination of urea in human and animal urine and in wine.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
95580
Xanthydrol, for the detection of urea, ≥99.0% (HPLC)
C13H10O2
252573
Xanthydrol, 98%
C13H10O2