EMAIL THIS PAGE TO A FRIEND
Applied optics
Femtosecond pulse shaping adds a new dimension to mass spectrometry.
PMID 17571144
Abstract
Phase-shaped femtosecond laser pulses and mass spectrometry were implemented as a tool for improving molecular identification. We demonstrate that the specific lines in the mass spectra of several chemical warfare simulants are sensitive to the phase characteristics of the incident laser field. The deviation in the relative yield of fragment ions observed upon pulse shaping (enhancement or suppression) adds a new dimension to mass spectrometry that improves molecular identification and can be used to quantitatively analyze mixtures of isomers.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
41430
Dimethyl phosphite, purum, ≥98.0% (GC)
C2H7O3P
D178454
Dimethyl phosphite, 98%
C2H7O3P