EMAIL THIS PAGE TO A FRIEND
Journal of applied physiology (Bethesda, Md. : 1985)
Thiol modification in H2O2- and thromboxane-induced vaso- and bronchoconstriction in rat perfused lung.
PMID 1757354
Abstract
Hydrogen peroxide (H2O2), arachidonic acid (AA), and U-44069, a thromboxane analogue, all induced vaso- and bronchoconstriction in the isolated perfused rat lung. The role of protein sulfhydryl modifications in these processes was investigated. The thiol oxidizing agent diamide inhibited both vaso- and bronchoconstriction induced by H2O2, AA, or U-44069. Diamide had only a marginal effect on glutathione and protein thiol levels and no effect on lung mechanics. The diamide inhibition was reversible, and H2O2-induced vaso- and bronchoconstriction was almost maximal after 10 min of perfusion with buffer. The recovery was more rapid if dithiothreitol, a thiol reducing agent, was used in the buffer. H2O2- and AA-induced vaso- and bronchoconstriction is caused by thromboxane release. Diamide did not influence H2O2- or AA-dependent thromboxane formation, indicating that neither AA release nor AA metabolism to thromboxane is sensitive to thiol oxidation. Thus our results indicate that the site of diamide-induced thiol oxidation is the thromboxane receptor or its signal transduction.